accessibility menu, dialog, popup

UserName

CAS RN 5332-96-7

CH3ONOO

CAS RN 5332-96-7

IUPAC Name: 1-(4-nitrophenyl)propan-2-one
Synonyms: 1-(4-nitrophenyl)propan-2-one
Molecular Weight (g/mol): 179.18
Molecular Formula: C9H9NO3
InChi Key: GEWWCWZGHNIUBW-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results