accessibility menu, dialog, popup

UserName

NH2HN

CAS RN 534-85-0

IUPAC Name: N1-phenylbenzene-1,2-diamine
Synonyms: 1,2-benzenediamine, N-phenyl-
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: NFCPRRWCTNLGSN-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1NC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)