missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-o-phenylenediamine, 98%

Product Code. 11362527
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362527 25 g 25g
11352527 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362527 Supplier Thermo Scientific Alfa Aesar Supplier No. L11616.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 534-85-0 | C12H12N2 | 184.24 g/mol

N-Phenyl-o-phenylenediamine is also used as an intermediate in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenyl-o-phenylenediamine is also used as an intermediate in organic synthesis and pharmaceuticals.

Solubility
Soluble in acetone, chloroform and benzene. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Store at room temperature. Incompatible with acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-85-0
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00007685
InChI Key NFCPRRWCTNLGSN-UHFFFAOYSA-N
Synonym 2-aminodiphenylamine, n-phenyl-o-phenylenediamine, n1-phenylbenzene-1,2-diamine, o-semidine, 1,2-benzenediamine, n-phenyl, o-aminodiphenylamine, n-phenyl-1,2-phenylenediamine, n-phenylbenzene-1,2-diamine, o-phenylenediamine, n-phenyl, unii-g2x60k1y26
PubChem CID 68297
IUPAC Name 2-N-phenylbenzene-1,2-diamine
SMILES NC1=CC=CC=C1NC1=CC=CC=C1

Specifications

Melting Point 77°C to 80°C
Quantity 25 g
Beilstein 908984
Solubility Information Soluble in acetone,chloroform and benzene. Insoluble in water.
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material N-Phenyl-o-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.