CAS RN 5340-78-3
IUPAC Name:
ethyl 3,3-dimethylbutanoate
Synonyms:
ethyl 3,3-dimethylbutanoate
Molecular Weight (g/mol):
144.21
Molecular Formula:
C8H16O2
InChi Key:
JWMNHAMYTBAUPI-UHFFFAOYSA-N
SMILES:
CCOC(=O)CC(C)(C)C
1
–
1
of
1
results
1
Ethyl tert-butylacetate, 98%
CAS: 5340-78-3 Molecular Formula: C8H16O2 Molecular Weight (g/mol): 144.214 MDL Number: MFCD00026911 InChI Key: JWMNHAMYTBAUPI-UHFFFAOYSA-N Synonym: ethyl tert-butylacetate,ethyl 3,3-dimethylbutyrate,t-c4h9ch2c o och2ch3,3,3-dimethylbutyric acid ethyl ester,jwmnhamytbaupi-uhfffaoysa,tert-butylacetic acid ethyl ester,tert-butylacetic ethyl ester,butanoic acid, 3,3-dimethyl-, ethyl ester PubChem CID: 79284 IUPAC Name: ethyl 3,3-dimethylbutanoate SMILES: CCOC(=O)CC(C)(C)C