missing translation for 'onlineSavingsMsg'
Learn More

Ethyl tert-butylacetate, 98%

Product Code. 11347505
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347505 100 g 100g
11327505 5 g 5g
11337505 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347505 Supplier Thermo Scientific Alfa Aesar Supplier No. L05603.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5340-78-3 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5340-78-3
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00026911
InChI Key JWMNHAMYTBAUPI-UHFFFAOYSA-N
Synonym ethyl tert-butylacetate, ethyl 3,3-dimethylbutyrate, t-c4h9ch2c o och2ch3, 3,3-dimethylbutyric acid ethyl ester, jwmnhamytbaupi-uhfffaoysa, tert-butylacetic acid ethyl ester, tert-butylacetic ethyl ester, butanoic acid, 3,3-dimethyl-, ethyl ester
PubChem CID 79284
IUPAC Name ethyl 3,3-dimethylbutanoate
SMILES CCOC(=O)CC(C)(C)C

Specifications

Density 0.86
Boiling Point 144°C to 146°C
Flash Point 34°C (93°F)
Refractive Index 1.4035
Quantity 100 g
UN Number UN3272
Beilstein 1749778
Formula Weight 144.22
Percent Purity 98%
Chemical Name or Material Ethyl tert-butylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.