CAS RN 5344-27-4
IUPAC Name:
2-(pyridin-4-yl)ethan-1-ol
Synonyms:
4-pyridineethanol
Molecular Weight (g/mol):
123.16
Molecular Formula:
C7H9NO
InChi Key:
DWPYQDGDWBKJQL-UHFFFAOYSA-N
SMILES:
OCCC1=CC=NC=C1
1
–
1
of
1
results
1
4-(2-Hydroxyethyl)pyridine, 98%
CAS: 5344-27-4 Molecular Formula: C7H9NO Molecular Weight (g/mol): 123.155 MDL Number: MFCD00129038 InChI Key: DWPYQDGDWBKJQL-UHFFFAOYSA-N Synonym: 4-pyridineethanol,2-pyridin-4-yl ethanol,4-2-hydroxyethyl pyridine,4-ethanolpyridine,2-4-pyridyl ethanol,2-pyridin-4-yl ethan-1-ol,2-pyridin-4-yl-ethanol,2-gamma-pyridyl ethanol,unii-i89rdq5yd9 PubChem CID: 72921 IUPAC Name: 2-pyridin-4-ylethanol SMILES: C1=CN=CC=C1CCO