missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)pyridine, 98%

Product Code. 11393237
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393237 1 g 1g
11303247 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393237 Supplier Thermo Scientific Alfa Aesar Supplier No. L13780.03
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5344-27-4 | C7H9NO | 123.155 g/mol

4-(2-Hydroxyethyl)pyridine acts as a reagent for the protection of carboxyl groups as their 2-(4-pyridyl)ethyl esters in organic synthesis. Further, it is used as an active pharmaceutical ingredient intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Hydroxyethyl)pyridine acts as a reagent for the protection of carboxyl groups as their 2-(4-pyridyl)ethyl esters in organic synthesis. Further, it is used as an active pharmaceutical ingredient intermediate.

Solubility
Miscible with water.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents, bases and mineral acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5344-27-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00129038
InChI Key DWPYQDGDWBKJQL-UHFFFAOYSA-N
Synonym 4-pyridineethanol, 2-pyridin-4-yl ethanol, 4-2-hydroxyethyl pyridine, 4-ethanolpyridine, 2-4-pyridyl ethanol, 2-pyridin-4-yl ethan-1-ol, 2-pyridin-4-yl-ethanol, 2-gamma-pyridyl ethanol, unii-i89rdq5yd9
PubChem CID 72921
IUPAC Name 2-pyridin-4-ylethanol
SMILES C1=CN=CC=C1CCO

Specifications

Melting Point 8°C to 10°C
Density 1.093
Boiling Point 121°C to 122°C (2 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.538
Quantity 1 g
Beilstein 111361
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 4-(2-Hydroxyethyl)pyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.