accessibility menu, dialog, popup

UserName

NH2ClClH2N

CAS RN 5348-42-5

IUPAC Name: 4,5-dichlorobenzene-1,2-diamine
Synonyms: 4,5-dichlorobenzene-1,2-diamine
Molecular Weight (g/mol): 177.03
Molecular Formula: C6H6Cl2N2
InChi Key: IWFHBRFJOHTIPU-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(Cl)C=C1N
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)