missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dichloro-o-phenylenediamine, 98%

Product Code. 10796691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10796691 25 g Glass bottle 25g
10713841 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10796691 Supplier Thermo Scientific Acros Supplier No. 113640250
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5348-42-5 | C6H6Cl2N2 | 177.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5348-42-5
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.03
MDL Number MFCD00007723
InChI Key IWFHBRFJOHTIPU-UHFFFAOYSA-N
Synonym 4,5-dichloro-o-phenylenediamine, 4,5-dichloro-1,2-benzenediamine, 1,2-diamino-4,5-dichlorobenzene, 4,5-dichloro-1,2-phenylenediamine, 1,2-benzenediamine, 4,5-dichloro, 4,5-dichloro-o-phenylendiamine, 1,2-dichloro-4,5-diaminobenzene, 2-amino-4,5-dichloroaniline, 4,5-dichloro-ortho-phenylenediamine, 4,5-dichlorophenylene-1,2-diamine
PubChem CID 79297
IUPAC Name 4,5-dichlorobenzene-1,2-diamine
SMILES NC1=CC(Cl)=C(Cl)C=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 162.0°C to 163.0°C
Color Brown to Brown-Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula Cl2C6H2(NH2)2
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 177.03
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4, 5-Dichloro-o-phenylenediamine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.