accessibility menu, dialog, popup

UserName

BrNOO

CAS RN 53534-14-8

IUPAC Name: 1-(3-bromophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(3-bromophenyl)pyrrole-2,5-dione
Molecular Weight (g/mol): 252.07
Molecular Formula: C10H6BrNO2
InChi Key: KJJYDECLTBAKCK-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)N1C(=O)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results