missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Bromophenyl)maleimide, 96%, Thermo Scientific™

Product Code. 11490060
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490060 250 mg 250mg
11400070 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490060 Supplier Thermo Scientific Alfa Aesar Supplier No. H50586.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-(3-Bromophenyl)maleimide is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53534-14-8
Molecular Formula C10H6BrNO2
Molecular Weight (g/mol) 252.067
MDL Number MFCD01026470
InChI Key KJJYDECLTBAKCK-UHFFFAOYSA-N
Synonym 1-3-bromophenyl-1h-pyrrole-2,5-dione, n-3-bromophenyl maleimide, 1-3-bromophenyl pyrrole-2,5-dione, 1-3-bromophenyl-2,5-dihydro-1h-pyrrole-2,5-dione, acmc-20apbw, cyto8b11, cbmicro_044659, 1-3-bromophenyl maleimide, 1-3-bromophenyl azoline-2,5-dione, 1-3-bromo-phenyl-pyrrole-2,5-dione
PubChem CID 708855
IUPAC Name 1-(3-bromophenyl)pyrrole-2,5-dione
SMILES C1=CC(=CC(=C1)Br)N2C(=O)C=CC2=O

Specifikationer

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 252.07
Percent Purity 96%
Chemical Name or Material N-(3-Bromophenyl)maleimide
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.