accessibility menu, dialog, popup

UserName

CAS RN 5355-42-0

NHN

CAS RN 5355-42-0

IUPAC Name: 3-[(piperidin-1-yl)methyl]-1H-indole
Synonyms: 3-(piperidin-1-ylmethyl)-1H-indole
Molecular Weight (g/mol): 214.31
Molecular Formula: C14H18N2
InChi Key: HPKXPFRRVVWAPD-UHFFFAOYSA-N
SMILES: C(N1CCCCC1)C1=CNC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results