missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Piperidinylmethyl)indole, 95%, Thermo Scientific™

Product Code. 15459858
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15459858 250 mg 250mg
15439858 1 g 1g
15449858 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 15459858

Brand: Thermo Scientific Alfa Aesar H63256.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(1-Piperidinylmethyl)indole, is used as a pharmaceutical intermediate.

Solubility
Freely soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5355-42-0
Molecular Formula C14H18N2
Molecular Weight (g/mol) 214.312
MDL Number MFCD00458412
InChI Key HPKXPFRRVVWAPD-UHFFFAOYSA-N
Synonym n-skatylpiperidine, 3-piperidin-1-yl methyl-1h-indole, 3-piperidinomethyl indole, indole, 3-piperidinomethyl, 3-piperidin-1-ylmethyl-1h-indole, chembl57535, 3-piperidinomethyl-1h-indole, 3-1-piperidinylmethyl-1h-indole, 3-piperidylmethyl indole
PubChem CID 21454
IUPAC Name 3-(piperidin-1-ylmethyl)-1H-indole
SMILES C1CCN(CC1)CC2=CNC3=CC=CC=C32

Specifications

Color Tan to White
Quantity 250 mg
Beilstein 176547
Solubility Information Freely soluble in water.
Formula Weight 214.3
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(1-Piperidinylmethyl)indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.