accessibility menu, dialog, popup

UserName

CH3ONCO

CAS RN 5395-71-1

IUPAC Name: 1-ethoxy-2-isocyanatobenzene
Synonyms: 1-ethoxy-2-isocyanatobenzene
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: BUIQXUQLYZPMLS-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results