missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyphenyl isocyanate, 97%

Product Code. 11387676
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11387676 5 g 5g
11397676 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387676 Supplier Thermo Scientific Alfa Aesar Supplier No. L11299.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5395-71-1 | C9H9NO2 | 163.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5395-71-1
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00002007
InChI Key BUIQXUQLYZPMLS-UHFFFAOYSA-N
Synonym 2-ethoxyphenyl isocyanate, o-ethoxyphenyl isocyanate, benzene, 1-ethoxy-2-isocyanato, isocyanic acid, o-ethoxyphenyl ester, 2-ethoxyphenylisocyanate, 2-ethoxybenzenisocyanate, acmc-20anwp, ethoxyphenyl isocyanate, 2-ethoxy phenyl isocyanate, #
PubChem CID 79374
IUPAC Name 1-ethoxy-2-isocyanatobenzene
SMILES CCOC1=CC=CC=C1N=C=O

Specifications

Density 1.105
Boiling Point 119°C to 120°C (20 mmHg)
Flash Point 102°C (215°F)
Refractive Index 1.527
Quantity 5 g
UN Number UN2206
Beilstein 1102956
Sensitivity Moisture sensitive
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 2-Ethoxyphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.