CAS RN 53985-48-1
IUPAC Name:
3-methoxy-5-(trifluoromethyl)benzoic acid
Synonyms:
3-methoxy-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
220.15
Molecular Formula:
C9H7F3O3
InChi Key:
VAQSHRZOMRFBCX-UHFFFAOYSA-N
SMILES:
COC1=CC(=CC(=C1)C(F)(F)F)C(O)=O
1
–
1
of
1
results
1
3-Methoxy-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™
CAS: 53985-48-1 Molecular Formula: C9H7F3O3 Molecular Weight (g/mol): 220.147 MDL Number: MFCD09025409 InChI Key: VAQSHRZOMRFBCX-UHFFFAOYSA-N Synonym: 3-methoxy-5-trifluoromethyl benzoic acid,3-methoxy-5-trifluoromethyl-benzoic acid,benzoic acid, 3-methoxy-5-trifluoromethyl,3-carboxy-5-methoxybenzotrifluoride,3-carboxy-5-trifluoromethyl anisole,3methoxy-5-trifluoromethylbenzoic acid,3-methoxy-5-trifluoromethyl benzoicacid,5-methoxy-3-trifluoromethyl benzoic acid,3-trifluoromethyl-5-methoxybenzoic acid PubChem CID: 21544467 IUPAC Name: 3-methoxy-5-(trifluoromethyl)benzoic acid SMILES: COC1=CC(=CC(=C1)C(F)(F)F)C(=O)O