accessibility menu, dialog, popup

UserName

KHOOSOHOO

CAS RN 5399-63-3

IUPAC Name: potassium 4-sulfobenzoic acid
Synonyms: potassium 4-sulfobenzoic acid
Molecular Weight (g/mol): 241.28
Molecular Formula: C7H6KO5S
InChi Key: PXRJBUPXKDXDLG-UHFFFAOYSA-N
SMILES: [K+].OC(=O)C1=CC=C(C=C1)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results