CAS RN 54-20-6
IUPAC Name:
5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms:
trifluoromethyluracil
Molecular Weight (g/mol):
180.09
Molecular Formula:
C5H3F3N2O2
InChi Key:
LMNPKIOZMGYQIU-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CNC(=O)NC1=O
1
–
2
of
2
results
1
Trifluorothymine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-(Trifluoromethyl)uracil, 97%
CAS: 54-20-6 Molecular Formula: C5H3F3N2O2 Molecular Weight (g/mol): 180.09 MDL Number: MFCD00006024 InChI Key: LMNPKIOZMGYQIU-UHFFFAOYSA-N Synonym: 5-trifluoromethyl uracil,5-trifluoromethyluracil,5-trifluoromethyl pyrimidine-2,4 1h,3h-dione,trifluoromethyluracil,2,4 1h,3h-pyrimidinedione, 5-trifluoromethyl,5-tfmu,uracil, 5-trifluoromethyl,5-trifluoromethyl pyrimidine-2,4-diol,5-trifluoromethyl-uracil PubChem CID: 5899 IUPAC Name: 5-(trifluoromethyl)-1H-pyrimidine-2,4-dione SMILES: FC(F)(F)C1=CNC(=O)NC1=O