missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)uracil, 97%

Product Code. 11382927
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382927 250 mg 250mg
11372927 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11382927 Supplier Thermo Scientific Alfa Aesar Supplier No. L16196.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 54-20-6 | C5H3F3N2O2 | 180.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-20-6
Molecular Formula C5H3F3N2O2
Molecular Weight (g/mol) 180.09
MDL Number MFCD00006024
InChI Key LMNPKIOZMGYQIU-UHFFFAOYSA-N
Synonym 5-trifluoromethyl uracil, 5-trifluoromethyluracil, 5-trifluoromethyl pyrimidine-2,4 1h,3h-dione, trifluoromethyluracil, 2,4 1h,3h-pyrimidinedione, 5-trifluoromethyl, 5-tfmu, uracil, 5-trifluoromethyl, 5-trifluoromethyl pyrimidine-2,4-diol, 5-trifluoromethyl-uracil
PubChem CID 5899
IUPAC Name 5-(trifluoromethyl)-1H-pyrimidine-2,4-dione
SMILES FC(F)(F)C1=CNC(=O)NC1=O

Specifications

Melting Point 251°C to 254
Quantity 250 mg
Beilstein 612694
Formula Weight 180.08
Percent Purity 97%
Chemical Name or Material 5-(Trifluoromethyl)uracil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.