accessibility menu, dialog, popup

UserName

H3COOCH3OH

CAS RN 5405-41-4

IUPAC Name: ethyl 3-hydroxybutanoate
Synonyms: ethyl 3-hydroxybutyrate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: OMSUIQOIVADKIM-UHFFFAOYNA-N
SMILES: CCOC(=O)CC(C)O
Quantity 
  • (2)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)