missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-hydroxybutyrate, 98+%

Product Code. 11437084
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437084 100 g 100g
11427084 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11437084 Supplier Thermo Scientific Alfa Aesar Supplier No. B21521.22
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5405-41-4 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5405-41-4
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00004545
InChI Key OMSUIQOIVADKIM-UHFFFAOYNA-N
Synonym ethyl 3-hydroxybutyrate, ethyl beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, ethyl ester, butyric acid, 3-hydroxy-, ethyl ester, fema no. 3428, ethyl 3-hydroxybutyrate natural, ethyl dl-3-hydroxybutyrate, ethyl 1-3-hydroxybutyrate, dl-3-hydroxy-n-butyric acid ethyl ester, ethyl3-hydroxybutyrate
PubChem CID 62572
ChEBI CHEBI:87685
IUPAC Name ethyl 3-hydroxybutanoate
SMILES CCOC(=O)CC(C)O

Specifications

Density 1.013
Boiling Point 169°C to 171°C
Flash Point 64°C (148°F)
Linear Formula CH3CHOHCH2CO2CH2CH3
Refractive Index 1.421
Quantity 100 g
Beilstein 1446190
Formula Weight 132.16
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-hydroxybutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.