CAS RN 54125-61-0
CAS RN 54125-61-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methoxycarbonyl-2-methyltetralone, TRC
CAS: 54125-61-0 Molecular Formula: C13H14O3 Molecular Weight (g/mol): 218.25 Synonym: Methyl 2-Methyl-1-tetralone-2-carboxylate,Methyl 1,2,3,4-Tetrahydro-2-methyl-1-oxo-2-naphthalenecarboxylate IUPAC Name: methyl 2-methyl-1-oxo-3,4-dihydronaphthalene-2-carboxylate SMILES: COC(=O)C1(C)CCc2ccccc2C1=O