CAS RN 5445-17-0
IUPAC Name:
methyl 2-bromopropanoate
Synonyms:
methyl 2-bromopropanoate
Molecular Weight (g/mol):
167
Molecular Formula:
C4H7BrO2
InChi Key:
ACEONLNNWKIPTM-UHFFFAOYNA-N
SMILES:
COC(=O)C(C)Br
1
–
4
of
4
results
1
Methyl DL-2-bromopropionate, 99%
CAS: 5445-17-0 Molecular Formula: C4H7BrO2 Molecular Weight (g/mol): 167 MDL Number: MFCD00000143 InChI Key: ACEONLNNWKIPTM-UHFFFAOYSA-N PubChem CID: 95576 IUPAC Name: methyl 2-bromopropanoate SMILES: CC(C(=O)OC)Br
2
Methyl 2-Bromopropionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methyl 2-Bromopropionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Methyl 2-Bromopropionate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.