missing translation for 'onlineSavingsMsg'
Learn More

Methyl DL-2-bromopropionate, 99%

Product Code. 10702221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10702221 100 g Glass bottle 100g
10397300 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10702221 Supplier Thermo Scientific Acros Supplier No. 173391000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5445-17-0 | C4H7BrO2 | 167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5445-17-0
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167
MDL Number MFCD00000143
InChI Key ACEONLNNWKIPTM-UHFFFAOYSA-N
PubChem CID 95576
IUPAC Name methyl 2-bromopropanoate
SMILES CC(C(=O)OC)Br

Specifications

Color Colorless to Yellow
Density 1.4970g/mL
Boiling Point 143.0°C to 145.0°C (19.0 mmHg)
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(Br)CO2CH3
Refractive Index 1.4509 to 1.4530
Quantity 100 g
Beilstein 02, 253
Specific Gravity 1.497
Synonym methyl 2-bromopropionate, propanoic acid, 2-bromo-, methyl ester, 2-bromopropionic acid methyl ester, methyl dl-2-bromopropionate, methyl2-bromopropionate, methyl alpha-bromopropionate, methyl .alpha.-bromopropionate, methyl-2-bromopropanoate, methyl 2-bromo-propionate, propionic acid, 2-bromo-, methyl ester
Solubility Information Solubility in water: immiscible
Formula Weight 167
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl DL-2-bromopropionate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.