accessibility menu, dialog, popup

UserName

CAS RN 54594-06-8

NH2H2COHO

CAS RN 54594-06-8

IUPAC Name: 2-aminopent-4-enoic acid
Synonyms: allylglycine
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: WNNNWFKQCKFSDK-UHFFFAOYNA-N
SMILES: NC(CC=C)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)