missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Allylglycine, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | D-Allylglycine |
| CAS | 54594-06-8 |
| Molecular Formula | C5 H9 N O2 |
| Quantity | 50 mg |
| Synonym | 4-Pentenoic acid, 2-amino-, (2R)-, 4-Pentenoic acid, 2-amino-, (R)-, (2R)-2-Amino-4-pentenoic acid, (+)-Allylglycine, (2R)-2-Amino-4-pentenoic acid, (R)-2-Amino-4-pentenoic acid, (R)-Allylglycine, (R)-α-Allylglycine, D-2-Amino-4-pentenoic acid, D-Allylglycine, D-2-Allylglycine, D-α-Allylglycine |
| InChI Formula | InChI=1S/C5H9NO2/c1-2-3-4(6)5(7)8/h2,4H,1,3,6H2,(H,7,8)/t4-/m1/s1 |
| SMILES | N[C@H](CC=C)C(=O)O |
| IUPAC Name | (2R)-2-aminopent-4-enoic acid |
| Molecular Weight (g/mol) | 115.13 |
| Formula Weight | 115.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction