accessibility menu, dialog, popup

UserName

CH3OOLi

CAS RN 546-89-4

IUPAC Name: lithium(1+) acetate
Synonyms: AcOLi quilonorm acetic acid, lithium salt acetic acid lithium salt lithium acetate (anh.) anhydrous lithium acetate lithium acetate (anhydrous) LiOAc
Molecular Weight (g/mol): 65.98
Molecular Formula: C2H3LiO2
InChi Key: XIXADJRWDQXREU-UHFFFAOYSA-M
SMILES: [Li+].CC([O-])=O
Grade 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)