missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate, 99+%, for analysis, anhydrous

Product Code. 11361143 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11361143 100 g Glass bottle 100g
10227370 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361143 Supplier Thermo Scientific Acros Supplier No. 297111000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 546-89-4 | C2H3LiO2 | 65.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-89-4
Molecular Formula C2H3LiO2
Molecular Weight (g/mol) 65.98
MDL Number MFCD00013057
InChI Key XIXADJRWDQXREU-UHFFFAOYSA-M
Synonym lithium acetate, lithium ethanate, quilone, acoli, acetic acid, lithium salt, lithium ethanoate, lithium 1+ ion acetate, lioac, unii-h4278o5flv, acetic acid lithium salt
PubChem CID 3474584
ChEBI CHEBI:63045
IUPAC Name lithium;acetate
SMILES [Li+].CC([O-])=O

Specifications

Melting Point 280.0°C to 285.0°C
Physical Form Solid
Quantity 100 g
Assay Percent Range 99+%
Linear Formula CH3COOLi
Beilstein 02,107
Merck Index 15,5577
Solubility Information Solubility in water: 408g/L (20°C). Other solubilities: 667g/L (40°C), 938g/L (50°C), freely soluble in alcohol
Formula Weight 65.99
Percent Purity ≥99%
Grade Analytical
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Lithium acetate
Show More Show Less
compliance-icons
Product Identifier
  • Lithium acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Acetic acid, lithiumsalt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.