accessibility menu, dialog, popup

UserName

CH3OOCH3HO

CAS RN 547-64-8

IUPAC Name: methyl 2-hydroxypropanoate
Synonyms: methyl lactate
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: LPEKGGXMPWTOCB-UHFFFAOYNA-N
SMILES: COC(=O)C(C)O
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results