missing translation for 'onlineSavingsMsg'
Learn More

Methyl DL-lactate, 99%

Product Code. 15426928
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426928 25 mL 25mL
15416928 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15426928 Supplier Thermo Scientific Alfa Aesar Supplier No. H60455.AC
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 547-64-8 | C4H8O3 | 104.105 g/mol

Methyl DL-lactate is used as a solvent for nitrocellulose, cellulase acetate, cellulase acetobutyrote and cellulase acetaprapionate. It is used in the manufacture of lacquers and dopes where it contributes high tolerance for diluents, good flaw and blush resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl DL-lactate is used as a solvent for nitrocellulose, cellulase acetate, cellulase acetobutyrote and cellulase acetaprapionate. It is used in the manufacture of lacquers and dopes where it contributes high tolerance for diluents, good flaw and blush resistance.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store away from moisture and strong bases. Do not store together with acids. Store under dry inert gas. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-64-8
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00066367
InChI Key LPEKGGXMPWTOCB-UHFFFAOYSA-N
Synonym methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate
PubChem CID 11040
ChEBI CHEBI:83221
IUPAC Name methyl 2-hydroxypropanoate
SMILES CC(C(=O)OC)O

Specifications

Density 1.093
Flash Point 51°C (124°F)
Refractive Index 1.413
Quantity 25 mL
UN Number UN3272
Beilstein 1720586
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 104.1
Percent Purity 99%
Chemical Name or Material Methyl DL-lactate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.