accessibility menu, dialog, popup

UserName

CAS RN 54997-90-9

H3CCH3SOOCl

CAS RN 54997-90-9

IUPAC Name: 4-(propan-2-yl)benzene-1-sulfonyl chloride
Synonyms: 4-isopropylbenzenesulfonyl chloride
Molecular Weight (g/mol): 218.7
Molecular Formula: C9H11ClO2S
InChi Key: CETRNHJIXGITKR-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)S(Cl)(=O)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results