accessibility menu, dialog, popup

UserName

OHHOO(R)OH(R)HO(R)HO

CAS RN 551-68-8

IUPAC Name: (3R,4R,5R)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms: d-psicose D-altrulose D-erythro-hexulose D-ribo-hex-2-ulose D-ribo-2-ketohexulose D-ribo-2-hexulose D-allulose D-pseudofructose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: LKDRXBCSQODPBY-JDJSBBGDSA-N
SMILES: OCC1(O)OC[C@@H](O)[C@@H](O)[C@H]1O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results