missing translation for 'onlineSavingsMsg'
Learn More

D-Psicose, 98%

Product Code. 15365068 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15365068 5 g 5g
15355068 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15365068 Supplier Thermo Scientific Acros Supplier No. 456270050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 551-68-8 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 551-68-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00083478
InChI Key BJHIKXHVCXFQLS-PUFIMZNGSA-N
Synonym erythrohexulose, 3r,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, pseudofructose, unii-qcc18lng3e, d-ribo-hex-2-ulose, qcc18lng3e, dl-psicose, ribo-2-hexulose, d-ribo-2-ketohexose, keto-d-psicose
PubChem CID 90008
ChEBI CHEBI:27605
IUPAC Name (3R,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 109.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol, insoluble in acetone
Formula Weight 180.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Psicose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.