accessibility menu, dialog, popup

UserName

CAS RN 5527-94-6

CH3ClF

CAS RN 5527-94-6

IUPAC Name: 1-chloro-2-fluoro-4-methylbenzene
Synonyms: 1-chloro-2-fluoro-4-methylbenzene
Molecular Weight (g/mol): 144.57
Molecular Formula: C7H6ClF
InChi Key: MHXNHUSVBYUTJL-UHFFFAOYSA-N
SMILES: CC1=CC(F)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results