accessibility menu, dialog, popup

UserName

CH3BrNOO

CAS RN 55289-35-5

IUPAC Name: 1-bromo-2-methyl-3-nitrobenzene
Synonyms: 1-bromo-2-methyl-3-nitrobenzene
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: LYTNSGFSAXWBCA-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1Br)[N+]([O-])=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)