accessibility menu, dialog, popup

UserName

CAS RN 55513-19-4

NNHF

CAS RN 55513-19-4

IUPAC Name: 1-[(3-fluorophenyl)methyl]piperazine
Synonyms: 1-[(3-fluorophenyl)methyl]piperazine
Molecular Weight (g/mol): 194.25
Molecular Formula: C11H15FN2
InChi Key: ITHBJSRWFNLKIH-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CN2CCNCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results