accessibility menu, dialog, popup

UserName

CAS RN 55737-66-1

CH3OOOHONOO

CAS RN 55737-66-1

IUPAC Name: 4-(methoxycarbonyl)-2-nitrobenzoic acid
Synonyms: 4-(methoxycarbonyl)-2-nitrobenzoic acid
Molecular Weight (g/mol): 225.16
Molecular Formula: C9H7NO6
InChi Key: VULISSQANNKDCH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=C(C=C1)C(O)=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results