CAS RN 55750-61-3
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)acetate
Synonyms:
2,5-dioxopyrrolidin-1-yl 2-(2,5-dioxopyrrol-1-yl)acetate
Molecular Weight (g/mol):
252.18
Molecular Formula:
C10H8N2O6
InChi Key:
TYKASZBHFXBROF-UHFFFAOYSA-N
SMILES:
O=C(CN1C(=O)C=CC1=O)ON1C(=O)CCC1=O