missing translation for 'onlineSavingsMsg'
Learn More

Maleimidoacetic Acid N-Hydroxysuccinimide Ester, TRC

Product Code. 30478505
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478505 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478505 Supplier Toronto Research Chemicals Supplier No. TRCM12500050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 55750-61-3
Molecular Formula C10 H8 N2 O6
Quantity 50 mg
InChI Formula InChI=1S/C10H8N2O6/c13-6-1-2-7(14)11(6)5-10(17)18-12-8(15)3-4-9(12)16/h1-2H,3-5H2
SMILES O=C(CN1C(=O)C=CC1=O)ON2C(=O)CCC2=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-(2,5-dioxopyrrol-1-yl)acetate
Molecular Weight (g/mol) 252.18
Formula Weight 252.0382
Chemical Name or Material Maleimidoacetic Acid N-Hydroxysuccinimide Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.