CAS RN 5588-16-9
CAS RN 5588-16-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Althiazide, TRC
CAS: 5588-16-9 Molecular Formula: C11 H14 Cl N3 O4 S3 Molecular Weight (g/mol): 383.89 Synonym: Altizide,(3RS)-6-Chloro-3-[(prop-2-enylsulfanyl)methyl]-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide IUPAC Name: 6-chloro-1,1-dioxo-3-(prop-2-enylsulfanylmethyl)-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide SMILES: NS(=O)(=O)c1cc2c(NC(CSCC=C)NS2(=O)=O)cc1Cl