missing translation for 'onlineSavingsMsg'
Learn More

Althiazide, TRC

Product Code. 30138602
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138602 1 mg 1mg
30006649 10 mg 10mg
30005490 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138602 Supplier Toronto Research Chemicals Supplier No. TRCA5757751MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5588-16-9
Molecular Formula C11 H14 Cl N3 O4 S3
Molecular Weight (g/mol) 383.89
Synonym Altizide, (3RS)-6-Chloro-3-[(prop-2-enylsulfanyl)methyl]-3,4-dihydro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide
IUPAC Name 6-chloro-1,1-dioxo-3-(prop-2-enylsulfanylmethyl)-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)c1cc2c(NC(CSCC=C)NS2(=O)=O)cc1Cl

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C11H14ClN3O4S3/c1-2-3-20-6-11-14-8-4-7(12)9(21(13,16)17)5-10(8)22(18,19)15-11/h2,4-5,11,14-15H,1,3,6H2,(H2,13,16,17)
Formula Weight 382.9835
Chemical Name or Material Altizide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.