CAS RN 5589-96-8
CAS RN 5589-96-8
IUPAC Name:
2-bromo-2-chloroacetic acid
Synonyms:
bromochloroacetic acid keratin
Molecular Weight (g/mol):
173.39
Molecular Formula:
C2H2BrClO2
InChi Key:
GEHJBWKLJVFKPS-UHFFFAOYNA-N
SMILES:
OC(=O)C(Cl)Br
1
–
3
of
3
results
1
Bromochloroacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bromochloroacetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.