accessibility menu, dialog, popup

UserName

ClCH3NH3CCH3

CAS RN 56-37-1

IUPAC Name: benzyltriethylazanium chloride
Synonyms: benzyl triethyl ammonium chloride
Molecular Weight (g/mol): 227.78
Molecular Formula: C13H22ClN
InChi Key: HTZCNXWZYVXIMZ-UHFFFAOYSA-M
SMILES: [Cl-].CC[N+](CC)(CC)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)