accessibility menu, dialog, popup

UserName

CAS RN 56096-89-0

FIOHO

CAS RN 56096-89-0

IUPAC Name: 4-fluoro-2-iodobenzoic acid
Synonyms: 4-fluoro-2-iodobenzoic acid
Molecular Weight (g/mol): 266.01
Molecular Formula: C7H4FIO2
InChi Key: DUKFTVLJAXWGPI-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(I)C=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results