accessibility menu, dialog, popup

UserName

RbCH3OO

CAS RN 563-67-7

IUPAC Name: rubidium(1+) acetate
Synonyms: rubidium(1+) acetate
Molecular Weight (g/mol): 144.51
Molecular Formula: C2H3O2Rb
InChi Key: FOGKDYADEBOSPL-UHFFFAOYSA-M
SMILES: [Rb+].CC([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results