missing translation for 'onlineSavingsMsg'
Learn More

Rubidium acetate, 99.8% (metals basis)

Product Code. 11337627
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337627 10 g 10g
11327627 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Codice prodotto. 11337627 Fornitore Thermo Scientific Alfa Aesar N. del fornitore 012890.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 563-67-7 | C2H3O2Rb | 144.51 g/mol

Rubidium acetate has been reported to modify the acid/base character of the catalyst supports like silica, alumina, and titania. It can be used in the preparation of nanosized rubidium ferrite, and as a promoter on the palladium-catalyzed synthesis of vinyl acetate in heterogeneous gas reaction. It is used in medical and in research laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 563-67-7
Molecular Formula C2H3O2Rb
Molecular Weight (g/mol) 144.51
MDL Number MFCD00013058
InChI Key FOGKDYADEBOSPL-UHFFFAOYSA-M
Synonym rubidium acetate, acetic acid, rubidium salt, unii-86h795sz6d, acetic acid, rubidium salt 1:1, acetic acid rubidium, rubidio acetate, acmc-1aks3, rubidium 1+ acetate ion, rubidium 1+ ion acetate, c2h3o2.rb
PubChem CID 23673628
IUPAC Name rubidium(1+);acetate
SMILES [Rb+].CC([O-])=O

Specifica

Melting Point 246°C
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
Formula Weight 144.52
Percent Purity 99.8%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Rubidium acetate

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.