accessibility menu, dialog, popup

UserName

CAS RN 565-61-7

IUPAC Name: 3-methylpentan-2-one
Synonyms: 3-methyl-2-pentanone
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: UIHCLUNTQKBZGK-UHFFFAOYNA-N
SMILES: CCC(C)C(C)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)