missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pentanone, 99%

Product Code. 11934821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11934821 25 g Glass bottle 25g
11914821 1 g Glass bottle 1g
11924821 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11934821 Supplier Thermo Scientific Acros Supplier No. 127380250
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 565-61-7 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-61-7
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
InChI Key UIHCLUNTQKBZGK-UHFFFAOYSA-N
Synonym 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
PubChem CID 11262
IUPAC Name 3-methylpentan-2-one
SMILES CCC(C)C(=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.8100g/mL
Boiling Point 118.0°C (758.0 mmHg)
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(CH3)COCH3
Refractive Index 1.4002
Quantity 25 g
Beilstein 01,693
Specific Gravity 0.81
Formula Weight 100.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-pentanone
Show More Show Less
compliance-icons
Product Identifier
  • 3-Methyl-2-pentanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P223-Do not allow contact with water.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.