accessibility menu, dialog, popup

UserName

H3CONH2OCH3OHO

CAS RN 5653-40-7

IUPAC Name: 2-amino-4,5-dimethoxybenzoic acid
Synonyms: 2-amino-4,5-dimethoxybenzoic acid
Molecular Weight (g/mol): 197.19
Molecular Formula: C9H11NO4
InChi Key: HJVAVGOPTDJYOJ-UHFFFAOYSA-N
SMILES: COC1=CC(N)=C(C=C1OC)C(O)=O
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results