accessibility menu, dialog, popup

UserName

CH3OOFFFFFFFFFFFFFFFFFFFFFFF

CAS RN 56554-52-0

IUPAC Name: methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate
Synonyms: methyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoate
Molecular Weight (g/mol): 628.13
Molecular Formula: C13H3F23O2
InChi Key: ABNZIVVYHQYSLI-UHFFFAOYSA-N
SMILES: COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results